Learn More
c-Kit Recombinant Rabbit Monoclonal Antibody (HC34LC14), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Supplier: Thermo Scientific 701494
Description
This antibody is predicted to react with bovine, equine, goat, non-human primate, rat and porcine based on sequence homology. Intact IgG appears on a non-reducing gel as ∽150 kDa band and upon reduction generating a ∽25 kDa light chain band and a ∽50 kDa heavy chain. Recombit rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombit rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
CD117 is a member of the tyrosine kinase receptor family and is expressed by subsets of hematopoietic progenitor cells and mast cells. This proto-oncogene binds to stem cell factor to form a dimer. This interaction promotes proliferation and differentiation of hematopoietic progenitor cells, and mast cell differentiation. CD117 is also expressed by melanocytes and plays a role in signaling and activation of these cells.Specifications
c-Kit | |
Recombinant Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
P10721 | |
KIT | |
Peptide corresponding to amino acids 246–261 of human c-Kit | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Western Blot | |
HC34LC14 | |
Unconjugated | |
KIT | |
C-Kit, CD117, PBT, SCFR, mast/stem cell growth factor receptor Kit, p145 c-kit, piebald trait protein, proto-oncogene c-Kit, proto-oncogene tyrosine-protein kinase Kit, soluble KIT variant 1, tyrosine-protein kinase Kit, v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene-like protein, ckit | |
Rabbit | |
Protein A | |
RUO | |
3815 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.