Learn More
C.L. Sturkey C.L. Sturkey Diamond Disposable Microtome Blades
Unique blade that has a special effectiveness on hard tissue and decalcified bone
Manufacturer:  C.L. Sturkey DT315DD
 View more versions of this product
Catalog No. 22-210003
Add to cart
Description
Description
Features an amorphous diamond coating that is applied to the cutting edge, resulting in a very tough and durable blade
Packaging Options
  • Bulk Dispenser: Holds 120 Heavy Duty High-Profile or 200 Low-Profile blades per dispenser
  • Box Packaging: Holds 100 Heavy Duty High-Profile or 170 Low-Profile blades per box (non-dispensing)
Specifications
Specifications
Low
Yes
Amorphous Diamond
200 Pack
Documents
Accessories