Learn More
C.L. Sturkey EXTREMUS Disposable Microtome Blades
Uniquely coated to suit the widest variety of cutting conditions
Supplier: C.L. Sturkey DT315X
Description
Offers a unique combination of durability and precision that saves time and money
Packaging Options
- Standard Dispenser: Holds 50 blades per dispenser
- Bulk Dispenser: Holds 120 Heavy Duty High-Profile or 200 Low-Profile blades per dispenser
- Box Packaging: Holds 100 Heavy Duty High-Profile or 170 Low-Profile blades per box (non-dispensing)
Specifications
Low | |
Yes | |
170 Pk. |