Learn More
C.L. Sturkey C.L. Sturkey Gold Disposable Microtome Blades
The ultimate blade for cutting hard tissue and bone
Manufacturer:  C.L. Sturkey DT315GD
 View more versions of this product
Catalog No. 22-210001
Add to cart
Description
Description
High-performance specialty blade, tough and durable
Packaging Options
  • Standard Dispenser: Holds 50 blades per dispenser
  • Bulk Dispenser: Holds 120 Heavy Duty High-Profile or 200 Low-Profile blades per dispenser
  • Box Packaging: Holds 100 Heavy Duty High-Profile or 170 Low-Profile blades per box (non-dispensing)
Specifications
Specifications
Low
Yes
Titanium Nitride
200 Pack
Documents
Accessories