C.L. Sturkey Permanent Microtome Knife
A premium line of permanent, steel microtome knives that have earned national and international recognition
Manufacturer: CL Sturkey K250D
 View more versions of this product
Catalog No. 22-210025
  Check Availability
Add to Cart
    Specifications
    Length (English) Blade 9.84 in.
    Length (Metric) Blade 25cm
    Type Permanent Microtome Knife
    Material Tempered Steel
    For Use With (Application) For general Paraffin and cryostat use
    Back Style Flat
  • Description & Specifications
Related Products
Product Content Feedback