Learn More
C.L. Sturkey Permanent Microtome Knife
A premium line of permanent, steel microtome knives that have earned national and international recognition
Manufacturer:  CL Sturkey K160A
 View more versions of this product
Catalog No. 22-210018
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Permanent Microtome Knife
For general Paraffin and cryostat use
6.29 in.
Tempered Steel
Round
16cm