Learn More
C.L. Sturkey C.L. Sturkey Premium Disposable Microtome Blades
Durable and economical uncoated blade
Manufacturer:  C.L. Sturkey D554PD
 View more versions of this product
Catalog No. 22-210007
Add to cart
Description
Description
Offered in heavy-duty High-Profile only
Packaging Options
  • Bulk Dispenser: Holds 120 High-Profile blades per dispenser
  • Box Packaging: Holds 100 High-Profile blades per box (non-dispensing)
Specifications
Specifications
High
Yes
120 Pack
Documents
Accessories