Learn More
C.L. Sturkey Reconditioning/Sharpening Service
Please note that we must receive your microtome knife in order to perform this service
Manufacturer:  C.L. Sturkey R250
 View more versions of this product
Catalog No. 22-210030
Add to cart
Description
Description
Permanent microtome knife reconditioning is a professional sharpening service that employs the same craftsmanship and expert care used to manufacture them. Knives of all sizes and styles from any manufacturer can be serviced. For information about about transportation of knives to C.L. Sturkey, please contact Fisher Scientific Customer Service.
Specifications
Specifications
250mm
Microtome Knife
Carbon Steel