Learn More
C.L. Sturkey C.L. Sturkey Silver Disposable Microtome Blades
Durable and long-lasting, ideal for general paraffin and cryostat use
Manufacturer:  C.L. Sturkey DT315RD
 View more versions of this product
Catalog No. 22-210000
Add to cart
Description
Description
Coated with a nitride and copolymer finish to provide smooth, wrinkle-free sections
Packaging Options
  • Standard Dispenser: Holds 50 blades per dispenser
  • Bulk Dispenser: Holds 200 Low-Profile blades per dispenser
  • Box Packaging: Holds 170 Low-Profile blades per box (non-dispensing)
Specifications
Specifications
Low
Yes
Nitride and Copolymer
200 Pack
Documents
Accessories