Learn More
c-MAF Monoclonal Antibody (sym0F1), eFluor™ 450, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$288.65
Specifications
Antigen | c-MAF |
---|---|
Clone | sym0F1 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
48-985-542
|
eBioscience
48985542 |
100 Tests |
Each of 1
for $288.65
Save 36% Reg : |
|
|||||
Description
The sym0F1 monoclonal antibody reacts with human and mouse c-Maf, a 42 kDa basic leucine zipper transcription factor shown to be involved in the neural, ocular and hematopoietic systems. In hematopoietic cells, it was first shown to be crucial for IL-4 ex
c-MAF Monoclonal antibody specifically detects c-MAF in Human, Mouse samples. It is validated for Flow Cytometry.Specifications
c-MAF | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
O75444, P54843 | |
IgG2b κ | |
4° C, store in dark, DO NOT FREEZE! | |
MAF |
sym0F1 | |
Flow Cytometry | |
eFluor 450 | |
Mouse | |
Human, Mouse | |
17132, 4094 | |
Primary | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.