Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

c-Maf Rabbit anti-Human, Mouse, Rat, Bovine, Alexa Fluor™ 350, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP224551AF350

Catalog No. NP2241A30


Add to cart

Description

Description

c-Maf Polyclonal antibody specifically detects c-Maf in Human,Mouse,Rat,Bovine samples. It is validated for Western Blot,Immunohistochemistry,Immunohistochemistry (Paraffin).
Specifications

Specifications

c-Maf
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
MAF
A synthetic peptide corresponding to amino acids 75-110 of human c-maf was used as the immunogen, GenBank no. NP_001026974.1 .
0.1 mL
Primary
4094
Bovine, Human, Mouse, Rat
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Avian musculoaponeurotic fibrosarcoma (MAF) protooncogene, c-MAF, c-maf proto-oncogene, MGC71685, Proto-oncogene c-Maf, T lymphocyte c-maf long form, transcription factor Maf, v-maf musculoaponeurotic fibrosarcoma (avian) oncogene homolog, V-maf musculoaponeurotic fibrosarcoma oncogene homolog, v-maf musculoaponeurotic fibrosarcoma oncogene homolog (avian)
Rabbit
IgG
Store at 4°C in the dark.
Polyclonal
100% homologous in human isoforms CRA_a, CRA_b, and CRA_c).
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit