Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

c-Maf Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$106.19 - $352.89

Specifications

Antigen c-Maf
Dilution Western Blot 1.0 ug/ml, Immunohistochemistry
Applications Western Blot, Immunohistochemistry
Conjugate Unconjugated
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB125863
View Documents
Novus Biologicals
NBP310481100UL
100 μg
Each for $352.89
Add to cart
 
NB125864
View Documents
Novus Biologicals
NBP31048125UL
25 μg
Each for $106.19
Add to cart
 
Description

Description

c-Maf Polyclonal antibody specifically detects c-Maf in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications

Specifications

c-Maf
Western Blot, Immunohistochemistry
Purified
RUO
PBS buffer, 2% sucrose
4094
Polyclonal
Affinity purified
Western Blot 1.0 ug/ml, Immunohistochemistry
Unconjugated
Rabbit
Human
Avian musculoaponeurotic fibrosarcoma (MAF) protooncogene, c-MAF, c-maf proto-oncogene, MGC71685, Proto-oncogene c-Maf, T lymphocyte c-maf long form, transcription factor Maf, v-maf musculoaponeurotic fibrosarcoma (avian) oncogene homolog, V-maf musculoaponeurotic fibrosarcoma oncogene homolog, v-maf musculoaponeurotic fibrosarcoma oncogene homolog (avian)
The immunogen is a synthetic peptide directed towards the N terminal region of human c-Maf (NP_001026974). Peptide sequence MASELAMSNSDLPTSPLAMEYVNDFDLMKFEVKKEPVETDRIISQCGRLI
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit