Learn More
c-Myb Mouse anti-Human, Alexa Fluor™ 750, Clone: MYB286, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP234690AF750
Catalog No. NP234690A70
Add to cart
Description
Description
c-Myb Monoclonal antibody specifically detects c-Myb in Human samples. It is validated for Western Blot,Flow Cytometry,ELISA,Immunocytochemistry,Immunofluorescence.
Specifications
Specifications
c-Myb
MYB286
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
c-myb, Cmyb, c-myb protein (140 AA), c-myb_CDS, c-myb10A_CDS, c-myb13A_CDS, c-myb14A_CDS, c-myb8B_CDS, efg, Proto-oncogene c-Myb, transcriptional activator Myb, v-myb avian myeloblastosis viral oncogene homolog, v-myb myeloblastosis viral oncogene homolog (avian)
Mouse
IgG1 Kappa
Store at 4°C in the dark.
Monoclonal
The highly leukemogenic avian retrovirus E26 contains two oncogenes, v-Myb and v-Ets, which are expressed together as a fusion protein. The cellular homolog of v-Myb, designated c-Myb, encodes a transcription factor. Deletion or disruption of a negative regulatory domain mapping within the carboxy-terminal domain of c-Myb results in enhanced transactivating capacity and in parallel, leads to activation of its ability to transform hemopoietic cells. c-Myb is expressed preferentially, but not exclusively, in immature hemopoietic cells and its expression decreases as cells differentiate.
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
Alexa Fluor 750
50 mM sodium borate with 0.05% sodium azide
MYB
A synthetic peptide, corresponding to aa 119-135 (RRKVEQEGYPQESSKAG) of c-myb oncoprotein, coupled to KLH
0.1 mL
Primary
4602
Human