Learn More
Thermo Scientific Lab Vision c-myb/v-myb Ab-1, Mouse Monoclonal Antibody
Ensure accurate and reproducible results in immunohistochemistry and western blotting experiments with Thermo Scientific c-myb/v-myb Ab-1, Mouse Monoclonal Antibody.
Manufacturer: Thermo Scientific MS1728R1
Description
Ensure accurate and reproducible results in immunohistochemistry and western blotting experiments with Thermo Scientific c-myb/v-myb Ab-1, Mouse Monoclonal Antibody.
Host Species: Mouse
Clone: 4E3
Isotype: IgG2a
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Recombinant v-myb protein
Molecular Weight: ~80 kDa
Positive Control: Jurkat and Ls174T cells, Testis
Cellular Localization: Cytoplasmic and nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
Note:
USA: RUO; Int'l: RUO
Specifications
c-myb/v-myb Ab-1 | |
Monoclonal | |
Unconjugated | |
Prediluted | |
Recombinant v-myb protein | |
1mL | |
Transcription Regulation | |
Primary | |
Human |
Immunohistochemistry (Paraffin), Western Blot | |
4E3 | |
Ready-to-use for IHC (UVLP) | |
Murine | |
IgG2a | |
RUO | |
Primary | |
Monoclonal |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.