Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ c-myb/v-myb Ab-1, Mouse Monoclonal Antibody

Ensure accurate and reproducible results in immunohistochemistry and western blotting experiments with Thermo Scientific™ c-myb/v-myb Ab-1, Mouse Monoclonal Antibody.

Manufacturer:  Thermo Scientific™ MS1728P0

 View more versions of this product

Catalog No. MS1728P0

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Ensure accurate and reproducible results in immunohistochemistry and western blotting experiments with Thermo Scientific™ c-myb/v-myb Ab-1, Mouse Monoclonal Antibody.

Transcription factor Myb is expressed in hematopoietic cells and immature gastroepithelial cells.

Host Species: Mouse

Clone: 4E3

Isotype: IgG2a

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Recombinant v-myb protein

Molecular Weight: ~80 kDa

Positive Control: Jurkat and Ls174T cells, Testis

Cellular Localization: Cytoplasmic and nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

c-myb/v-myb Ab-1
Monoclonal
200μg/mL
Purified Ab with BSA and Azide
Murine
IgG2a
RUO
Primary
Monoclonal
Immunohistochemistry (Paraffin), Western Blot
4E3
Unconjugated
Concentrated
Recombinant v-myb protein
100μL
Transcription Regulation
Primary
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit