Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ c-myc Ab-2, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, flow cytometry and electron microscopy applications.

$199.10 - $780.50

Specifications

Antigen c-myc Ab-2
Clone 9E10.3
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Host Species Murine
View More Specs

Products 10
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
MS139B
View Documents
Thermo Scientific™
MS139B
Biotinylated Ab with BSA and Azide 1mL
Each for $780.50
Add to cart
 
MS139B0
View Documents
Thermo Scientific™
MS139B0
Biotinylated Ab with BSA and Azide 100μL
Each for $208.00
Add to cart
 
MS139B1
View Documents
Thermo Scientific™
MS139B1
Biotinylated Ab with BSA and Azide 500μL
Each for $677.50
Add to cart
 
MS139P0
View Documents
Thermo Scientific™
MS139P0
Purified Ab with BSA and Azide 100μL
Each for $199.10
Add to cart
 
MS139P1
View Documents
Thermo Scientific™
MS139P1
Purified Ab with BSA and Azide 500μL
Each for $422.00
Add to cart
 
MS139P1ABX
View Documents
Thermo Scientific™
MS139P1ABX
Ab without BSA/Azide (Frozen) 100μL
Each for $473.50
Add to cart
 
MS139PABX
View Documents
Thermo Scientific™
MS139PABX
Ab without BSA/Azide (Frozen) 200μL
Each for $639.00
Add to cart
 
MS139R7
View Documents
Thermo Scientific™
MS139R7
Ready-to-use for IHC (UVLP) 7mL
Each for $413.00
Add to cart
 
MS139P
View Documents
Thermo Scientific™
MS139P
Purified Ab with BSA and Azide 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
MS139R1
View Documents
Thermo Scientific™
MS139R1
Ready-to-use for IHC (UVLP) 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description

Description

Ensure accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, flow cytometry and electron microscopy applications with Thermo Scientific™ c-myc Ab-2, Mouse Monoclonal Antibody.

c-myc is involved in the control of cell proliferation and differentiation and is amplified and/or overexpressed in a variety of tumors.

Host Species: Mouse

Clone: 9E10.3

Isotype: IgG1/κ

Species Reactivity: Human. Does not react with mouse, rat and chicken. Others not tested.

Epitope: aa 410-419

Immunogen: A synthetic peptide, aa 408-439, (AEEQKLISEEDLLRKRREQLKHKLEQLRNSCA) from C-terminus of human c-myc

Molecular Weight: 64-67 kDa

Positive Control: Raji cells, Breast CA

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Flow Cytometry
  • Electron Microscopy
  • Affinity Purification, Immunoprecipitation and Western Blotting of fusion proteins labeled with EQKLISEEDL tag

Note:

USA: RUO; Int'l: RUO

<
Specifications

Specifications

c-myc Ab-2
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Murine
Transcription Regulation
A synthetic peptide, aa 408-439, (AEEQKLISEEDLLRKRREQLKHKLEQLRNSCA) from C-terminus of human c-myc
9E10.3
Monoclonal
RUO
Primary
IgG2a kappa
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit