Learn More
c-Myc Mouse anti-Human, Primate, Mouse, Alexa Fluor™ 750, Clone: MYC699, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP234692AF750
Catalog No. NP234692A70
Add to cart
Description
Description
c-Myc Monoclonal antibody specifically detects c-Myc in Human,Primate,Mouse samples. It is validated for Flow Cytometry,Immunocytochemistry,Immunofluorescence.
Specifications
Specifications
c-Myc
MYC699
Flow Cytometry, Immunocytochemistry/Immunofluorescence
avian myelocytomatosis viral oncogene homolog, BHLHE39, bHLHe39MRTL, Class E basic helix-loop-helix protein 39, c-Myc, MYC, myc proto-oncogene protein, MYCC, myc-related translation/localization regulatory factor, Proto-oncogene c-Myc, Transcription factor p64, v-myc avian myelocytomatosis viral oncogene homolog, v-myc myelocytomatosis viral oncogene homolog (avian)
Mouse
IgG1 Kappa
0.1 mL
Store at 4°C in the dark.
Monoclonal
The c-Myc protein is a transcription factor, which is encoded by the c-Myc gene on human chromosome 8q24. c-Myc is commonly activated in a variety of tumor cells and plays an important role in cellular proliferation, differentiation, apoptosis and cell cycle progression. The phosphorylation of c-Myc has been investigated and previous studies have suggested a functional association between phosphorylation at Thr58/Ser62 by glycogen synthase kinase 3, cyclin dependent kinase, ERK2 and C-Jun N terminal Kinase (JNK) in cell proliferation and cell cycle regulation. Studies also have shown that c-Myc is essential for tumor cell development in vasculogenesis and angiogenesis that distribute blood throughout the cells, and which brought extensive attention in the development of new therapeutic approach for cancer treatment.
Flow Cytometry, Immunocytochemistry, Immunofluorescence
Alexa Fluor 750
50 mM sodium borate with 0.05% sodium azide
MYC
A synthetic peptide, corresponding to aa 408-439 (AEEQKLISEEDLLRKRREQLKHKLEQL-RNSCA) from C-terminus of human c-Myc Antibody (MYC699), coupled to KLH
63 kDa
Autophagy, Cancer, Cancer Stem Cells, Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, Epigenetics, Epitope Tags, Stem Cell Markers, Transcription Factors and Regulators, Tumor Suppressors
Primary
4609
Human, Primate, Mouse