Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
c-Myc Mouse anti-Non-species specific, Alexa Fluor™ 532, Clone: Myc.A7, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP237822AF532
Description
c-Myc Monoclonal specifically detects c-Myc in Non-species specific samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
c-Myc | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
MYC | |
Synthetic peptide: EQKLISEEDL c-Myc Antibody (Myc.A7). | |
0.1 mL | |
Primary | |
Detects N-terminal or C-terminal Myc tagged fusion proteins. It has been successfully used in ELISA, immunofluorescence, immunoprecipitation and Western Blot procedures. immunogen is a synthetic peptide EQKLISEEDL (Myc). | |
IgG1 |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
Myc.A7 | |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence | |
avian myelocytomatosis viral oncogene homolog, BHLHE39, bHLHe39MRTL, Class E basic helix-loop-helix protein 39, c-Myc, MYC, myc proto-oncogene protein, MYCC, myc-related translation/localization regulatory factor, Proto-oncogene c-Myc, Transcription factor p64, v-myc avian myelocytomatosis viral oncogene homolog, v-myc myelocytomatosis viral oncogene homolog (avian) | |
Mouse | |
63 kDa | |
Autophagy, Cancer, Cancer Stem Cells, Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, Epigenetics, Epitope Tags, Stem Cell Markers, Transcription Factors and Regulators, Tumor Suppressors | |
4609 | |
Nonspecies specific |
Safety and Handling
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title