Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

c-Myc Rabbit anti-Human,Mouse, Unconjugated, Clone: 7E18, MilliporeSigma™

Rabbit Monoclonal Antibody

Specifications

Antigen c-Myc
Applications Immunocytochemistry, Immunohistochemistry, Knockdown, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ZRB100325MI
View Documents
MilliporeSigma
ZRB100325UL
25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
ZRB10034XMI
View Documents
MilliporeSigma
ZRB10034X25UL
4 x 25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

c-Myc Monoclonal antibody specifically detects c-Myc in Human,Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry, RNAi/siRNA/Gene Knockdown, Western Blotting.
Specifications

Specifications

c-Myc
Unconjugated
Rabbit
Myc proto-oncogene protein;Class E basic helix-loop-helix protein 39;bHLHe39;Proto-oncogene c-Myc;Transcription factor p64
GST/His-tagged recombinant fragment corresponding to 109 amino acids from the internal region of human c-Myc.
Primary
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
Immunocytochemistry, Immunohistochemistry, Knockdown, Western Blot
Purified
Human, Mouse
MYC, BHLHE39
Monoclonal
Protein A Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit