Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
c-Myc-responsive protein Rcl Rabbit anti-Human, Mouse, Clone: 7C1Z8, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | c-Myc-responsive protein Rcl |
---|---|
Clone | 7C1Z8 |
Applications | Western Blot, Immunofluorescence |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
c-Myc-responsive protein Rcl Monoclonal antibody specifically detects c-Myc-responsive protein Rcl in Human, Mouse samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
c-Myc-responsive protein Rcl | |
Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Cell Biology, Cell Cycle and Replication | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
10591 | |
IgG | |
Affinity purified |
7C1Z8 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
chromosome 6 open reading frame 108, c-Myc-responsive protein Rcl, deoxyribonucleoside 5'-monophosphate N-glycosidase, dJ330M21.3, EC 3.2.2, EC 3.2.2.-, putative c-Myc-responsive, rcl, RP3-330M21.3 | |
A synthetic peptide corresponding to a sequence within amino acids 75-174 of human c-Myc-responsive protein Rcl (O43598). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title