Learn More
Aat BioquestSupplier Diversity Partner C-PC [C-Phycocyanin] *CAS 11016-15-2*  DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 2553
C-Phycocyanin (C-PC) occurs as the major phycobiliprotein in many cyanobacteria and as a secondary phycobiliprotein in some red algae.
Catalog No. 50-195-3182