Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C-Peptide Mouse anti-Human, Biotin, Clone: 1489, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP242830B

Catalog No. NBP242830B

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

C-Peptide
1489
ELISA
Purified
INS
Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human insulin. The exact sequence is proprietary.
Protein A purified
Diabetes Research, Lipid and Metabolism
Primary
3630
ELISA
Biotin
PBS with 0.05% Sodium Azide
IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin
Mouse
IgG2a
0.1 ml
Store at 4C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit