Learn More
C-Peptide Mouse anti-Human, DyLight 488, Clone: 1519, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP242831G
Catalog No. NBP242831G
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
C-Peptide
1519
ELISA
Purified
INS
Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human insulin. The exact sequence is proprietary.
Protein A purified
Diabetes Research, Lipid and Metabolism
Primary
3630
ELISA
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin
Mouse
IgG2b
0.1 ml
Store at 4C in the dark.
Monoclonal
Human