Learn More
C-Peptide Mouse anti-Human, HRP, Clone: 1519, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP242831H
Catalog No. NBP242831H
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
C-Peptide
1519
ELISA
Purified
INS
Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human insulin. The exact sequence is proprietary.
Protein A purified
Diabetes Research, Lipid and Metabolism
Primary
3630
ELISA
HRP
PBS with No Preservative
IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin
Mouse
IgG2b
0.1 ml
Store at 4C in the dark.
Monoclonal
Human