Learn More
C-Peptide Mouse, Clone: CC34, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Supplier:  Novus Biologicals NBP105433SS
 View more versions of this product
Catalog No. NBP105433SS
Add to cart
Description
Description
C-Peptide Monoclonal specifically detects C-Peptide in Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Frozen, Immunoassay, Sandwich ELISA.
Specifications
Specifications
C-Peptide
Monoclonal
Unconjugated
PBS (pH 7.4) with 0.09% Sodium Azide
INS
Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with fragments of rat C-peptides I and II conjugated with the carrier protein.
0.05 mg
Diabetes Research
3630
Mouse, Rat
Purified
ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoassay
CC34
ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, Immunoassay, Sandwich ELISA 1:100 - 1:2000
IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin
Mouse
Protein A purified
RUO
Primary
Reacts with rat c-peptide I, rat c-peptide II, mouse c-peptide II and mouse c-peptide I. Does not react with rat proinsulin.
Store at 4C. Do not freeze.
IgG1