Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C-Peptide Mouse, DyLight 405, Clone: CC34, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP105433V
Description
C-Peptide Monoclonal specifically detects C-Peptide in Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Frozen, Sandwich ELISA.Specifications
C-Peptide | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
INS | |
Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with fragments of rat C-peptides I and II conjugated with the carrier protein. | |
0.2 mL | |
Diabetes Research | |
3630 | |
Mouse, Rat | |
Purified |
ELISA, Immunohistochemistry, Immunohistochemistry (Frozen) | |
CC34 | |
ELISA, Immunohistochemistry, Immunohistochemistry-Frozen, Sandwich ELISA | |
IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Reacts with rat c-peptide I, rat c-peptide II, mouse c-peptide II and mouse c-peptide I. Does not react with rat proinsulin. | |
Store at 4C in the dark. | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title