Learn More
C-Peptide Mouse, HRP, Clone: 2B7, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NB1102555H
Catalog No. NB1102555H
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
C-Peptide Monoclonal antibody specifically detects Antigen in Bovine (Negative), Human, Mouse (Negative), Porcine (Negative), Rat (Negative) samples. It is validated for Enzyme Linked Immunosorbent Assay.
Specifications
Specifications
C-Peptide
2B7
ELISA
Purified
INS
Hybridoma clone has been derived from hybridization of X63-Ag8-653 myeloma cells with spleen cells of Balb/c mice immunized with recombinant C-peptide construction.
Protein A purified
RUO
Store at 4C in the dark.
Monoclonal
Affinity (Kd) > 1 x 10^8. Binds to the human free C-peptide and C-peptide region in proinsulin molecules. Antigenic determinants identified by MAbs are in the three-dimensional structure generated by N- and C- terminal regions of C-peptide separated by beta-turn at position 47-50 of proinsulin.. No cross-reactivity with human, bovine, porcine and mouse/rat insulin.
ELISA
HRP
PBS with No Preservative
IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin
Mouse
IgG1 Kappa
0.2 ml
Diabetes Research
Primary
3630
Human, Bovine (Negative), Mouse (Negative), Porcine (Negative), Rat (Negative)