Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C-Peptide Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$176.85 - $378.00
Specifications
Antigen | C-Peptide |
---|---|
Concentration | 0.5 mg/ml |
Dilution | Western Blot 1:200 |
Applications | Western Blot |
Classification | Polyclonal |
Description
C-Peptide Polyclonal specifically detects C-Peptide in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
C-Peptide | |
Western Blot 1:200 | |
Polyclonal | |
Rabbit | |
Diabetes Research | |
PBS (pH 7.2), 0.01% BSA, 50% Glycerol with 0.01% Thimerosal | |
3630 | |
Synthetic peptide (BV-L26) | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
0.5 mg/ml | |
Western Blot | |
Unconjugated | |
RUO | |
Human, Mouse, Rat | |
IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin | |
INS | |
IgG | |
Affinity Purified |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title