Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ C-Reactive Protein/CRP Antibody Pair [HRP]
Polyclonal/Monoclonal Antibody Pair
$386.00 - $832.00
Specifications
Assay Range | 34.38 pg/mL |
---|---|
Assay Sensitivity | 34.38-2200 pg/mL |
Assay | Sandwich ELISA |
Conjugate | HRP |
Immunoassay Kit Format | Sandwich ELISA Kit |
Description
This solid phase sandwich ELISA utilizes an antibody specific for Mouse C-Reactive Protein/CRP.Specifications
34.38 pg/mL | |
Sandwich ELISA | |
Sandwich ELISA Kit | |
This solid phase sandwich ELISA utilizes an antibody specific for Mouse C-Reactive Protein/CRP. | |
Sandwich ELISA | |
1401 | |
Rabbit | |
RUO | |
Storage is content dependent. | |
1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.25 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 2200 pg/mL is recommended. |
34.38-2200 pg/mL | |
HRP | |
Antibody Pairs | |
Mouse | |
C-reactive protein, C-reactive protein, pentraxin-related, MGC88244, pentraxin 1, PTX1MGC149895 | |
CRP | |
Cancer, Cytokine Research, Immunology, Innate Immunity | |
C-Reactive Protein/CRP |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title