Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C-Reactive Protein/CRP Rabbit anti-Human, Alexa Fluor 350, Clone: 106, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP289860AF350

Catalog No. NB068148


Add to cart

Description

Description

C-Reactive Protein/CRP Monoclonal antibody specifically detects C-Reactive Protein/CRP in Human samples. It is validated for ELISA
Specifications

Specifications

C-Reactive Protein/CRP
106
Sandwich ELISA Detection
Purified
Rabbit
IgG
0.1 mL
Cancer, Cytokine Research, Immunology, Innate Immunity
Primary
1401
ELISA
Alexa Fluor 350
50mM Sodium Borate
C-reactive protein, C-reactive protein, pentraxin-related, MGC88244, pentraxin 1, PTX1MGC149895
Recombinant Human C-Reactive Protein/CRP Protein
Protein A purified
RUO
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit