Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Alfa Aesar™ C-Type Natriuretic Peptide (32-53), Human
For cellular and molecular biology applications
Manufacturer:  Alfa Aesar™ J66540MCR
 View more versions of this product
Catalog No. AAJ66540MCR
Order in Bulk
Request bulk or custom formats
Specifications
Specifications
2197.59
Human
Solid
RUO
CNP-22
HPLC
1mg
Gly-Leu-Ser-Lys-Gly-Cys-Phe-Gly-Leu-Lys-Leu-Asp-Arg-Ile-Gly-Ser-Met-Ser-Gly-Leu-Gly-Cys (Disulfide bridge Cys6-Cys22)
-30° to -10°C
C-Type Natriuretic Peptide (32-53), human