Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C10orf18 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 204201AP150UL
Description
FAM208B is a protein coding gene.Specifications
C10orf18 | |
Polyclonal | |
Unconjugated | |
FAM208B | |
C10orf18, KIAA2006 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
54906 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.14 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q5VWN6 | |
FAM208B | |
C10orf18 Fusion Protein Ag14240 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction