Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VISTA Mouse anti-Human, Clone: CL3981, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA524704

Catalog No. PIMA524704


Add to cart

Description

Description

Immunogen sequence: LTFQDLHLHHG GHQAANTSHD LAQRHGLESA SDHHGNFSIT MRNLTLLDSG LYCCLVVEIR HHHSEHRVHG AMELQVQTGK DAPSNCVVYP SSS Binds to an epitope located within the peptide sequence LHLHHGGHQA as determined by overlapping synthetic peptides. Highest antigen sequence identity to the following orthologs - mouse 60%, rat 59%.

Immunoregulatory receptor which inhibits the T-cell response. May promote differentiation of embryonic stem cells, by inhibiting BMP4 signaling. May stimulate MMP14-mediated MMP2 activation.
Specifications

Specifications

VISTA
Monoclonal
1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9H7M9
SISP1; VISTA
Mouse
IgG1
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
64115
Immunohistochemistry
CL3981
Unconjugated
VSIR
Liquid
VSIR
Recombit Human C10orf54
Protein A
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only