Learn More
Aviva Systems Biology Corp C10orf56 Antibody - middle region (ARP55587_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP55587_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C10orf56. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human C10orf56. Alternate names: FLJ90798, C10orf56ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61547