Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM213A Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548580
Description
Predicted to react with bovine based on sequence homology.
Involved in redox regulation of the cell. Acts as an antioxidant. Inhibits TNFSF11-induced NFKB1 and JUN activation and osteoclast differentiation. May affect bone resorption and help to maintain bone mass.Specifications
FAM213A | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q9BRX8 | |
FAM213A | |
KLH conjugated synthetic peptide between 181-209 amino acids from the C-terminal region of human C10orf58. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
FAM213A | |
Redox-regulatory protein FAM213A, Peroxiredoxin-like 2 activated in M-CSF stimulated monocytes, Protein PAMM, FAM213A, C10orf58, PAMM | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84293 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction