Learn More
C10orf58 Rabbit anti-Human, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 204461AP
Catalog No. 50-173-2895
Add to cart
Description
Description
Involved in redox regulation of the cell. Acts as an antioxidant. Inhibits TNFSF11-induced NFKB1 and JUN activation and osteoclast differentiation. May affect bone resorption and help to maintain bone mass.
Specifications
Specifications
C10orf58
0.26 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q6AXX6, Q9BRX8
C10orf58, F213A, FAM213A, PSEC0139, UNQ611/PRO1198, UPF0765 protein C10orf58
Rabbit
IgG
150 μL
-20°C
Primary
361118, 84293
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
FAM213A
Liquid
Fam213a
C10orf58 Fusion Protein Ag14272
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Rat