Learn More
Aviva Systems Biology Corp C10orf83 Antibody - middle region (ARP53380_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP53380_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C10orf83. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human C10orf83. Alternate names: FLJ25925, RP11-548K23.4, bA548K23.4, C10orf83ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61012