Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C11orf20 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548709

Catalog No. PA548709

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with bovine based on sequence homology.

May be involved in late spermatogenesis.
Specifications

Specifications

C11orf20
Polyclonal
PBS with 0.09% sodium azide
Q9NTU4
Testis-expressed sequence 40 protein, TEX40, C11orf20
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
25858
Western Blot
Unconjugated
CATSPERZ
Liquid
CATSPERZ
KLH conjugated synthetic peptide between 172-200 amino acids from the C-terminal region of human C11orf20.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit