Learn More
C12orf54 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 251771AP
Catalog No. 50-173-4770
Add to cart
Specifications
Specifications
C12orf54
0.38 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q6X4T0
C12orf54, HSD 29, HSD 30, HSD30
Rabbit
IgG
150 μL
-20°C
Primary
121273
Immunohistochemistry (Paraffin)
Unconjugated
C12orf54
Liquid
C12orf54
C12orf54 Fusion Protein Ag15026
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human