Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C13orf15 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566622
Description
Immunogen sequence: LSDALCEFDAV LADFASPFHE RHFHYEEHLE RMK Highest antigen sequence identity to the following orthologs - mouse 94%, rat 94%.
Modulates the activity of cell cycle-specific kinases. Enhances CDK1 activity. May contribute to the regulation of the cell cycle. May inhibit growth of glioma cells by promoting arrest of mitotic progression at the G2/M transition. Fibrogenic factor contributing to the pathogenesis of renal fibrosis through fibroblast activation.
Specifications
C13orf15 | |
Polyclonal | |
Unconjugated | |
RGCC | |
1190002H23Rik; bA157L14.2; C13orf15; regulator of cell cycle; regulator of cell cycle RGCC; Response gene to complement 32 protein; Rgc32; RGC-32; Rgcc | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
28984 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H4X1 | |
RGCC | |
Recombinant Human C13orf15. Recombinant protein control fragment (Product #RP-107280). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction