Learn More
Aviva Systems Biology Corp C13orf30 Antibody - N-terminal region (ARP53425_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP53425_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C13orf30. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human C13orf30. Alternate names: FLJ40919, MGC138442, C13orf30ApplicationWB. Species reactivity: Human, Zebrafish
Catalog No. 50-014-63032