Learn More
C14orf143 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 198211AP
Catalog No. 50-173-2558
Add to cart
Description
Description
EFCAB11 gene ontology annotations related to this gene include calcium-mediated signaling; microtubule nucleation; mitotic spindle pole body duplication; spindle pole body duplication.
Specifications
Specifications
C14orf143
0.17 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9BUY7
C14orf143, EFCAB11
Rabbit
IgG
150 μL
-20°C
Primary
90141
Western Blot
Unconjugated
EFCAB11
Liquid
EFCAB11
C14orf143 Fusion Protein Ag13871
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human