Learn More
Aviva Systems Biology Corp RTRAF Antibody - N-terminal region (ARP34545_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP34545_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C14ORF166. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human C14orf166. Alternate names: CGI-99, CGI99, CLE, CLE7, LCRP369, RLLM1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-52964