Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C14orf166 Mouse anti-Human, Clone: OTI1E5, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA526642

Catalog No. PIMA526642


Add to cart

Description

Description

The function of the C14orf266 gene has not yet been determined.
Specifications

Specifications

C14orf166
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9Y224
RTRAF, C14orf166, CGI-99, CGI99, CLE, CLE7, LCRP369, RLLM1, hCLE1
Mouse
IgG2b
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
51637
Immunohistochemistry (Paraffin), Western Blot
OTI1E5
Unconjugated
C14ORF166
Liquid
C14ORF166
Full length human recombit protein of C14orf166 produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit