Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C14orf180 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP15968020UL

 View more versions of this product

Catalog No. NBP15968020


Add to cart

Description

Description

C14orf180 Polyclonal specifically detects C14orf180 in Human samples. It is validated for Western Blot.
Specifications

Specifications

C14orf180
Polyclonal
Western Blot 1:100-1:2000
Q8N912
C14ORF180
Synthetic peptides corresponding to C14ORF180 The peptide sequence was selected from the N terminal of C14ORF180. Peptide sequence RTAAGAVSPDSRPETRRQTRKNEEAAWGPRVCRAEREDNRKCPPSILKRS.
Affinity Purified
RUO
400258
Store at -20C. Avoid freeze-thaw cycles.
Western Blot
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
C14orf77, chromosome 14 open reading frame 180, chromosome 14 open reading frame 77, FLJ38594, NRAC, transmembrane protein C14orf180
Rabbit
18 kDa
20 μL
Primary
Human
IgG
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit