Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C14orf50 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549663

Catalog No. PA549663

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Inhibits phosphatase activity of protein phosphatase 1 (PP1) complexes.
Specifications

Specifications

C14orf50
Polyclonal
PBS with 0.09% sodium azide
Q96LQ0
Protein phosphatase 1 regulatory subunit 36, PPP1R36, C14orf50
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
145376
Western Blot
Unconjugated
PPP1R36
Liquid
PPP1R36
KLH conjugated synthetic peptide between 20-47 amino acids from the N-terminal region of human C14orf50.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit