Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C14orf50 Mouse anti-Human, Clone: OTI7A9, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527157

Catalog No. PIMA527157


Add to cart

Description

Description

Inhibits phosphatase activity of protein phosphatase 1 (PP1) complexes.
Specifications

Specifications

C14orf50
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q96LQ0
Protein phosphatase 1 regulatory subunit 36, PPP1R36, C14orf50
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
145376
Western Blot
OTI7A9
Unconjugated
PPP1R36
Liquid
PPP1R36
Full length human recombit protein of PPP1R36 produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit