Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C14orf68 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA518648
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
This gene encodes a member of a large family of mitochondrial transporters. The nuclear-encoded carrier protein is embedded in the inner mitochondrial membrane. This member of the family is thought to be an uncoupling protein that uncouples mitochondrial respiration from ATP synthesis by dissipating the transmembrane proton gradient. Alternative splicing results in multiple transcript variants.
Specifications
C14orf68 | |
Polyclonal | |
Unconjugated | |
SLC25A47 | |
C14orf68; HCC-down-regulated mitochondrial carrier protein; HDMCP; Hepatocellular carcinoma down-regulated mitochondrial carrier protein; hepatocellular carcinoma-downregulated mitochondrial carrier protein; HMFN1655; SLC25A47; Solute carrier family 25 member 47; solute carrier family 25, member 47 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
283600 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q6Q0C1 | |
SLC25A47 | |
Synthetic peptide sequence (KSRLQADGQGQRR) corresponding to the internal amino acids of C14orf68 (aa 242-255). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction