Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C14orf80 Mouse anti-Human, Clone: OTI2D6, liquid, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene TA811443

Description
C14orf80 may play a role in counteracting perturbation of actin filaments, such as after treatment with the actin depolymerizing microbial metabolite Chivosazole F.Specifications
C14orf80 | |
Monoclonal | |
Unconjugated | |
TEDC1 | |
C14orf80 | |
Mouse | |
Affinity Chromatography | |
RUO | |
283643 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
OTI2D6 | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q86SX3 | |
TEDC1 | |
Full length human recombit protein of human C14ORF80 produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title