Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C14orf80 Mouse anti-Human, Clone: OTI1H5, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527487

Catalog No. PIMA527487


Add to cart

Description

Description

C14orf80 may play a role in counteracting perturbation of actin filaments, such as after treatment with the actin depolymerizing microbial metabolite Chivosazole F.
Specifications

Specifications

C14orf80
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q86SX3
TEDC1, C14orf80
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
283643
Immunohistochemistry (Paraffin), Western Blot
OTI1H5
Unconjugated
C14ORF80
Liquid
C14ORF80
Full length human recombit protein of C14ORF80 produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit